Hooke'S Law Spring, Set of 5
470320-748
- Description:Hooke's law spring, set of 5
- Cat. no.:470320-748
Complete set of springs for investigating Hooke's law.
- Easy to read pointer for measuring elongation
A set of five springs with different spring constants. The hanging mass needed to produce 1 cm extension are 2.5 g, 5 g, 10 g, 15 g and 25 g respectively. Pre-stress has been removed for accurate results. With a hook to support a weight hanger and a red arrow to indicate position.
: Included manual.