Compound Bar Set
Compound bar set
Catalog # 470320-740
Supplier: United Scientific Supplies
Supplier #: BMSET04
Jump to:
Product Details & Documents
Thermal expansion of different metals.
- Four different metal combinations
- Quick and easy tools for demonstrating thermal expansion
There are four bar combinations: Brass/Aluminum, Brass/Iron, Copper/Iron, and Brass/ Copper. Since each metal has a different coefficient of thermal expansion, they will create a unique curve when paired with a different metal.
Metal bars are stamped with the identification of each metal, Aluminum (Al), Iron (Fe), Copper (Cu). The Brass bar is not stamped.
Delivery information: Includes activity guide.
Spécifications
- Désignation:Compound bar set