Cryogenic Lamination Tapes | Avantor
Cryogenic Lamination Tapes
Cryogenic Lamination Tapes
Certificates
Cryogenic Lamination Tapes
Cryogenic Lamination Tapes
3 Options Available Below
Product Details & Documents
Thin cryo lamination tape provides added protection against abrasion, harsh chemicals, and extreme temperatures. Designed to withstand long-term storage in liquid nitrogen and ultra-low temperature freezers, multiple freeze-thaw cycles, and transport on dry ice.
  • Temperature range: −196 to +93 °C (−321 to +199 °F)
  • Clear lamination tape protects material from extreme cryogenic conditions, including liquid phase and vapor phase liquid nitrogen
  • Resists harsh laboratory environments, including multiple freeze-thaw cycles
  • Offers protection against chemicals and solvents
  • Resistant to gamma irradiation (tested up to 50 KGy)
This ultra-thin cryo laminating tape provides excellent protection against most chemicals and reagents. It has also been designed to withstand long-term storage in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers (as low as 196 °C) when applied on cryo vials and tubes. Resistant to dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Core Size
    Dimensions
    Availability
    Price
  • Cryogenic lamination tape
    Transparent
    3"
    19×60000 mm (0.75×2400")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $68.34
    $68.34
    Roll of 1
       $68.34
    Supplier Number: 807656005867
  • Cryogenic lamination tape
    Transparent
    3"
    25.4×60000 mm (1×2400")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $87.56
    $87.56
    Roll of 1
       $87.56
    Supplier Number: 807656005874
  • Cryogenic lamination tape
    Transparent
    3"
    50.8×60000 mm (2×2400")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $108.00
    $108.00
    Roll of 1
       $108.00
    Supplier Number: 807656005881