Self-Laminating Calibration Labels | Avantor
Self-Laminating Calibration Labels
Self-Laminating Calibration Labels
Certificates
Self-Laminating Calibration Labels
Self-Laminating Calibration Labels
5 Options Available Below
Product Details & Documents
Self-laminating calibration labels for secure and safe identification of regularly inspected equipment and instruments. These labels can be inscribed with all relevant information and then laminated with the supplied clear laminate to provide physical protection against various abuses.
  • Temperature range: −80 to +93 °C (−112 to +200 °F)
  • Can withstand extreme temperatures, including cryogenic and high heat conditions
  • Resists abrasion and exposure to harsh chemicals and alcohols
  • Ideal for use with equipment that will undergo preventative maintenance, calibration, or re-certification
  • Once laminated the information is protected and cannot be altered
Self-laminating, writable, calibration labels designed for equipment that is calibrated or inspected regularly. The labels can be inscribed using permanent ink markers. The clear laminate is applied over the label, protecting the written information. These labels can resist abrasion and exposure to harsh chemicals, such as ethanol, acetone, isopropanol, thinners, detergents, and surface sanitizing wipes.
The self-laminating calibration labels are ideal for labeling regularly inspected medical equipment, equipment that has undergone preventative maintenance, that require calibration prior to use, or that are out of order and must not be used. Reagent calibration labels are also available for time-sensitive solutions prepared in the lab. Calibration label customization is also available to include the company's logos and information.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Legend
    Color
    Core Size
    Dimensions
    Packaging
    Availability
    Price
  • Self-laminating calibration labels
    Plastic
    Calibration
    3"
    19.1×38.1 mm (0.75×1.5")
    Discontinued
    Supplier Number: -
  • Self-laminating calibration labels
    Plastic
    Calibration
    Red on white
    3"
    25.4×54 mm (1×2.125")
    1000 labels/roll (1 label across)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $190.95
    $190.95
    Roll of 1
       $190.95
    Supplier Number: 807656040387
  • Self-laminating calibration labels
    Plastic
    Black on white
    3"
    25.4×54 mm (1×2.125")
    1000 labels/roll (1 label across)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $194.63
    $194.63
    Roll of 1
       $194.63
    Supplier Number: 807656110936
  • Self-laminating calibration labels
    Plastic
    Opened
    Black on green
    3"
    38.1×63.5 mm (1.5×2.5")
    500 labels/roll (1 label across)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $184.10
    $184.10
    Roll of 1
       $184.10
    Supplier Number: 807656040479
  • Self-laminating calibration labels
    Plastic
    Solution/Reagent
    Black on white
    3"
    63.5×101.6 mm (2.5×4")
    250 labels/roll (1 label across)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $211.67
    $211.67
    Roll of 1
       $211.67
    Supplier Number: 807656118734
  • Self-laminating calibration labels
    Plastic
    White on green
    3"
    88.9×50.8 mm (3.5×2")
    250 labels/roll (1 label across)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 250
    Roll of 250 - $148.85
    $148.85
    Roll of 250
       $148.85
    Supplier Number: 807656032955
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.