Single Channel Pipettes, Variable Volume, Labsciences
Supplier: LAB SCIENCE CA
Certificates
About this item
These pipettes have an ergonomic, lightweight design with a soft plunger movement to minimize hand fatigue and RSI.
- Simply turn the plunger for quick and easy volume setting
- Click stop digital adjustable volume mechanism
- Semi-autoclavable
- Uses Industry Standard pipette tips
- Color coded by volumes for easy identification
- Two stage plunger operation for 'blow-out' step during pipeting
The streamlined tip ejector allows easy access to narrow containers/tubes.
Certifications: Individually calibrated in accordance with DIN 12650 Standards and supplied with a computer generated calibration report.
Delivery: Includes a calibration tool for easy calibration and service.
Frequently Bought Together