Single Channel Pipettes, Fixed Volume, Labsciences
These pipettes have an ergonomic, lightweight design with a soft plunger movement to minimize hand fatigue and RSI.
- Semi-autoclavable
- Uses Industry Standard pipette tips
- Color coded by volumes for easy identification
- Two stage plunger operation for 'blow-out' step during pipetting
The streamlined tip ejector allows easy access to narrow containers/tubes.
: Individually calibrated in accordance with DIN 12650 Standards and supplied with a computer generated calibration report.
: Includes a mountable pipette holder for easy storage and a calibration tool for easy calibration and service.
Frequently Bought Together