Pipette Tips, Standard, Labsciences
Fournisseur: LAB SCIENCE CA
Certificats
À propos de cet article
These pipette tips are precision molded from virgin PP.
- Fully autoclavable in steam at 121 °C for 15 minutes
- Chemically resistant
- Contamination free
Tested for Fit and Function with most widely used pipettes on the market.
Frequently Bought Together