DYMO-Compatible Paper Labels
DYMO-Compatible Paper Labels
DYMO-Compatible Paper Labels
DYMO-Compatible Paper Labels
DYMO-Compatible Paper Labels
DYMO Compatible labels for printing with the DYMO 450-series. Available in assorted colors, they are ideal for color-coding and other general use applications in the lab and office.
  • Suitable for general use laboratory and office applications
  • Compatible with the 450 series and 4XL DYMO® LabelWriters
DYMO compatible paper labels for general use applications and laboratory organization. Ideal for office use on file folders, notebooks, storage boxes, paperwork, as well as shipping labels. Not suitable for deep-freeze applications.
Product Family Options
Rectangle permanent DYMO compatible paper labels
  • Désignation
    Matière
    Color
    Core Size
    Dimensions
    Packaging
  • DYMO compatible paper label
    Paper
    White
    1"
    12.7×25.4 mm (0.5×1")
    1000 labels/roll (2 labels across)
    Roll of 1
    Roll of 1 - $40.50
    $40.50
    Roll of 1
       $40.50
     : 807656006222
  • DYMO compatible paper label
    Paper
    Red
    1"
    12.7×25.4 mm (0.5×1")
    1000 labels/roll (2 labels across)
    Roll of 1
    Roll of 1 - $40.87
    $40.87
    Roll of 1
       $40.87
     : 807656024578
  • DYMO compatible paper label
    Paper
    Blue
    1"
    12.7×25.4 mm (0.5×1")
    1000 labels/roll (2 labels across)
    Roll of 1
    Roll of 1 - $40.50
    $40.50
    Roll of 1
       $40.50
     : 807656006840
  • DYMO compatible paper label
    Paper
    Bright green
    1"
    12.7×25.4 mm (0.5×1")
    1000 labels/roll (2 labels across)
    Roll of 1
    Roll of 1 - $40.50
    $40.50
    Roll of 1
       $40.50
     : 807656006215
  • DYMO compatible paper label
    Paper
    Yellow
    1"
    12.7×25.4 mm (0.5×1")
    1000 labels/roll (2 labels across)
    Roll of 1
    Roll of 1 - $40.50
    $40.50
    Roll of 1
       $40.50
     : 807656024585
  • DYMO compatible paper label
    Paper
    White
    1"
    14×87 mm (0.56×3.44")
    350 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $27.12
    $27.12
    Roll of 1
       $27.12
     : 807656024592
  • DYMO compatible paper label
    Paper
    White
    1"
    20×45 mm (0.79×1.77")
    500 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $46.19
    $46.19
    Roll of 1
       $46.19
     : 807656006000
  • DYMO compatible paper label
    Paper
    White
    1"
    25.4×25.4 mm (1×1")
    750 labels/roll (1 label across)
    Roll of 750
    Roll of 750 - $39.16
    $39.16
    Roll of 750
       $39.16
     : 807656062211
  • DYMO compatible paper label
    Paper
    White
    1"
    25.4×54 mm (1×2.12")
    500 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $51.53
    $51.53
    Roll of 1
       $51.53
     : 807656006413
  • DYMO compatible paper label
    Paper
    Yellow
    1"
    25.4×54 mm (1×2.12")
    500 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $51.53
    $51.53
    Roll of 1
       $51.53
     : 807656006420
  • DYMO compatible paper label
    Paper
    Blue
    1"
    25.4×54 mm (1×2.12")
    500 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $51.53
    $51.53
    Roll of 1
       $51.53
     : 807656006444
  • DYMO compatible paper label
    Paper
    Red
    1"
    25.4×54 mm (1×2.12")
    500 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $51.53
    $51.53
    Roll of 1
       $51.53
     : 807656006451
  • DYMO compatible paper label
    Paper
    Bright green
    1"
    25.4×54 mm (1×2.12")
    500 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $51.53
    $51.53
    Roll of 1
       $51.53
     : 807656006802
  • DYMO compatible paper label
    Paper
    Orange
    1"
    25.4×54 mm (1×2.12")
    500 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $51.53
    $51.53
    Roll of 1
       $51.53
     : 807656006437
  • DYMO compatible paper label
    Paper
    Pink
    1"
    25.4×54 mm (1×2.12")
    500 labels/roll (1 label across)
    Roll of 500
    Roll of 500 - $39.16
    $39.16
    Roll of 500
       $39.16
     : 807656062181
  • DYMO compatible paper label
    Paper
    White
    1"
    29×89 mm (1.12×3.5")
    350 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $32.53
    $32.53
    Roll of 1
       $32.53
     : 807656024608
  • DYMO compatible paper label
    Paper
    Yellow
    1"
    29×89 mm (1.12×3.5")
    350 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $32.53
    $32.53
    Roll of 1
       $32.53
     : 807656024615
  • DYMO compatible paper label
    Paper
    Blue
    1"
    29×89 mm (1.12×3.5")
    350 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $32.53
    $32.53
    Roll of 1
       $32.53
     : 807656024622
  • DYMO compatible paper label
    Paper
    Red
    1"
    29×89 mm (1.12×3.5")
    350 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $32.53
    $32.53
    Roll of 1
       $32.53
     : 807656024639
  • DYMO compatible paper label
    Paper
    Bright green
    1"
    29×89 mm (1.12×3.5")
    350 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $32.53
    $32.53
    Roll of 1
       $32.53
     : 807656024646
  • DYMO compatible paper label
    Paper
    Orange
    1"
    29×89 mm (1.12×3.5")
    350 labels/roll (1 label across)
    Roll of 1
    Roll of 1 - $32.53
    $32.53
    Roll of 1
       $32.53
     : 807656024653
  • DYMO compatible paper label
    Paper
    White
    1"
    35.56×88.9 mm (1.4×3.5")
    260 labels/roll (1 label across)
    Roll of 260
    Roll of 260 - $26.79
    $26.79
    Roll of 260
       $26.79
     : 807656083537
  • DYMO compatible paper label
    Paper
    Yellow
    1"
    35.56×88.9 mm (1.4×3.5")
    260 labels/roll (1 label across)
    Roll of 260
    Roll of 260 - $26.79
    $26.79
    Roll of 260
       $26.79
     : 807656083544
  • DYMO compatible paper label
    Paper
    Bright green
    1"
    35.56×88.9 mm (1.4×3.5")
    260 labels/roll (1 label across)
    Roll of 260
    Roll of 260 - $26.79
    $26.79
    Roll of 260
       $26.79
     : 807656083551
  • DYMO compatible paper label
    Paper
    White
    1"
    54×101 mm (2.12×3.98")
    300 labels/roll (1 label across)
    Roll of 3,000
    Roll of 3,000 - $53.61
    $53.61
    Roll of 3,000
       $53.61
     : 807656032931
  • DYMO compatible paper label
    Paper
    White
    1"
    57×32 mm (2.25×1.25")
    1000 labels/roll (1 label across)
     : 807656032948
Rectangle and circle permanent DYMO compatible paper labels
  • Désignation
    Matière
    Color
    Core Size
    Dimensions
    Packaging
  • DYMO compatible paper label
    Paper
    Yellow
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.37")
    500 labels/roll (1 set: rectangle+circle)
    Roll of 500
    Roll of 500 - $39.16
    $39.16
    Roll of 500
       $39.16
     : 807656006239
  • DYMO compatible paper label
    Paper
    White
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.37")
    500 labels/roll (1 set: rectangle+circle)
    Roll of 500
    Roll of 500 - $39.16
    $39.16
    Roll of 500
       $39.16
     : 807656006246
  • DYMO compatible paper label
    Paper
    Red
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.37")
    500 labels/roll (1 set: rectangle+circle)
    Roll of 500
    Roll of 500 - $39.16
    $39.16
    Roll of 500
       $39.16
     : 807656033495
  • DYMO compatible paper label
    Paper
    Dark brown
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.37")
    500 labels/roll (1 set: rectangle+circle)
    Roll of 500
    Roll of 500 - $39.16
    $39.16
    Roll of 500
       $39.16
     : 807656062228
  • DYMO compatible paper label
    Paper
    Bright green
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.37")
    500 labels/roll (1 set: rectangle+circle)
    Roll of 500
    Roll of 500 - $39.16
    $39.16
    Roll of 500
       $39.16
     : 807656062235
  • DYMO compatible paper label
    Paper
    Grey
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.37")
    500 labels/roll (1 set: rectangle+circle)
    Roll of 500
    Roll of 500 - $39.16
    $39.16
    Roll of 500
       $39.16
     : 807656062242
  • DYMO compatible paper label
    Paper
    Violet
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.37")
    500 labels/roll (1 set: rectangle+circle)
    Roll of 500
    Roll of 500 - $39.16
    $39.16
    Roll of 500
       $39.16
     : 807656062266
  • DYMO compatible paper label
    Paper
    Beige
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.37")
    500 labels/roll (1 set: rectangle+circle)
    Roll of 500
    Roll of 500 - $39.16
    $39.16
    Roll of 500
       $39.16
     : 807656999555