Vacuum Traps, Spherical Joints, Chemscience
With 35/20 spherical joints for added flexibility.
- Trap body has 50 mm O.D. and 360 mm length
: Order the three adapters and 35/20 SS pinch clamps separately.
With 35/20 spherical joints for added flexibility.
- Trap body has 50 mm O.D. and 360 mm length
: Order the three adapters and 35/20 SS pinch clamps separately.
Product Family Options
- DésignationST Joint No.
- : CS-V0175036
Specifications
Cat. No.CA10204-510 - : CS-V0175037
Specifications
Cat. No.CA10203-018 - : CS-V0175038
Specifications
Cat. No.CA10203-020 - : CS-V0175039
Specifications
Cat. No.CA10203-022 - : CS-CS610-35
Specifications
Cat. No.CA10203-024