Stopcocks, PTFE, Four Way, Chemscience
Supplier: CHEM SCIENCE INC CA
Jump to:
À propos de cet article
A 1:5 taper PTFE plug having double 'V', 90° bores to connect opposite sets of adjacent rams.
Configuration of stopcock allows it to be used in a system to obtain reversal of flow.
12 Options Available Below
Product Details & Documents
A 1:5 taper PTFE plug having double 'V', 90° bores to connect opposite sets of adjacent rams.
Configuration of stopcock allows it to be used in a system to obtain reversal of flow.
Product Family Options
Informations sur le produit
- DésignationStem O.D.Bore SizePlug SizeAvailabilityPrice
- Supplier Number: CS-S0421530Spécifications:
More
Specifications
Cat. No.CA10202-978 - Supplier Number: CS-S0341750Spécifications:
More
Specifications
Cat. No.CA10202-980 - Supplier Number: CS-S0342565Spécifications:
More
Specifications
Cat. No.CA10202-982 - Supplier Number: CS-S0701240Spécifications:
More
Specifications
Cat. No.CA10202-986 - Supplier Number: CS-S0701754Spécifications:
More
Specifications
Cat. No.CA10202-988 - Supplier Number: CS-S0701756Spécifications:
More
Specifications
Cat. No.CA10202-990 - Supplier Number: CS-S0711240Spécifications:
More
Specifications
Cat. No.CA10202-992 - Supplier Number: CS-S0711754Spécifications:
More
Specifications
Cat. No.CA10202-994 - Supplier Number: CS-S0711756Spécifications:
More
Specifications
Cat. No.CA10202-996 - Supplier Number: CS-S0721240Spécifications:
More
Specifications
Cat. No.CA10202-998 - Supplier Number: CS-S0721754Spécifications:
More
Specifications
Cat. No.CA10203-000 - Supplier Number: CS-S0721756Spécifications:
More
Specifications
Cat. No.CA10203-002