Stopper, Cap Design, Chemscience | Avantor
Stopper, Cap Design, Chemscience
Supplier:   CHEM SCIENCE INC CA
Stopper, Cap Design, Chemscience
Certificates
Stopper, Cap Design, Chemscience
Stopper, Cap Design, Chemscience
3 Options Available Below
Product Details & Documents
These stoppers are designed for apparatus having an inner opening.
Product Family Options
Informations sur le produit
  • Désignation
    ST Joint No.
    Availability
    Price
  • Stopper, cap design
    14/20
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $36.49
    $36.49
    Case of 1
       $36.49
    Supplier Number: CS-S0391420
  • Stopper, cap design
    19/22
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $56.79
    $56.79
    Case of 1
       $56.79
    Supplier Number: CS-S0391922
  • Stopper, cap design
    24/40
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $70.46
    $70.46
    Case of 1
       $70.46
    Supplier Number: CS-S0392440