Peptide Synthesis Vessels, Solid Phase, with Lower Joint and T-Bore PTFE Plug, Chemscience
These solid phase peptide synthesis vessels feature 2 mm PTFE stopcocks and fritted glass supports for resin beads during operation.
- 8 mm hose connection for vacuum
- Stopcock has an 8 mm O.D. side arm for inert gas introduction
The 24/40 inner drip joint can be assembled with other standard glassware.
: Vessels are supplied with PTFE-faced silicone lined black Nylon® caps.
Frequently Bought Together