Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience | Avantor
Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
Supplier:   CHEM SCIENCE INC CA
Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
Certificates
Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
8 Options Available Below
Product Details & Documents
With a fritted disc of coarse or medium porosity to support resin.
  • Top of the vessel has GL thread and a PTFE lined screw cap
  • PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
A connection of 8 mm O.D. is sealed on the inner joint for connection to an inner gas.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    GL Thread Size
    Fritted Disc Size
    Porosité
    Availability
    Price
  • 10 ml
    Peptide synthesis vessel, solid phase, T-bore, vacuum
    14
    15 mm
    Coarse
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $378.51
    $378.51
    Case of 1
       $378.51
    Supplier Number: CS-P0140010C
  • 10 ml
    Peptide synthesis vessel, solid phase, T-bore, vacuum
    14
    15 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $378.51
    $378.51
    Case of 1
       $378.51
    Supplier Number: CS-P0140010M
  • 25 ml
    Peptide synthesis vessel, solid phase, T-bore, vacuum
    25
    20 mm
    Coarse
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $414.45
    $414.45
    Case of 1
       $414.45
    Supplier Number: CS-P0140025C
  • 25 ml
    Peptide synthesis vessel, solid phase, T-bore, vacuum
    25
    20 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $421.37
    $421.37
    Case of 1
       $421.37
    Supplier Number: CS-P0140025M
  • 50 ml
    Peptide synthesis vessel, solid phase, T-bore, vacuum
    25
    25 mm
    Coarse
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $458.30
    $458.30
    Case of 1
       $458.30
    Supplier Number: CS-P0140050C
  • 50 ml
    Peptide synthesis vessel, solid phase, T-bore, vacuum
    25
    25 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $458.30
    $458.30
    Case of 1
       $458.30
    Supplier Number: CS-P0140050M
  • 100 ml
    Peptide synthesis vessel, solid phase, T-bore, vacuum
    25
    40 mm
    Coarse
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $497.20
    $497.20
    Case of 1
       $497.20
    Supplier Number: CS-P0140100C
  • 100 ml
    Peptide synthesis vessel, solid phase, T-bore, vacuum
    25
    40 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $497.20
    $497.20
    Case of 1
       $497.20
    Supplier Number: CS-P0140100M