BD BBL™ SIM Prepared Tube Media, BD Diagnostics
CA90001-180
: BD
:
- Pack type:Tube
- Description:SIM media
- Size:10/pack
- Cat. no.:CA90001-180
- Supplier No.:221010
The SIM prepared tube media is used to differentiate enteric bacilli on the basis of sulfide production, indole formation and motility.
Tube size K.