Peptide Synthesis Vessels, Solid Phase, T-Bore PTFE Stopcock, Chemscience | Avantor
Peptide Synthesis Vessels, Solid Phase, T-Bore PTFE Stopcock, Chemscience
Supplier:   CHEM SCIENCE INC CA
Peptide Synthesis Vessels, Solid Phase, T-Bore PTFE Stopcock, Chemscience
Certificates
Peptide Synthesis Vessels, Solid Phase, T-Bore PTFE Stopcock, Chemscience
Peptide Synthesis Vessels, Solid Phase, T-Bore PTFE Stopcock, Chemscience
9 Options Available Below
Product Details & Documents
With a fritted disc of coarse or medium porosity to support resin.
  • Top of the vessel has GL thread and a PTFE lined screw cap
  • PTFE stopcock has a 2 mm bore with the arm having 8 mm O.D.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    GL Thread Size
    Fritted Disc Size
    Porosité
    Availability
    Price
  • 10 ml
    Peptide synthesis vessel, solid phase, T-bore PTFE stopcock
    14
    15 mm
    Coarse
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $328.93
    $328.93
    Case of 1
       $328.93
    Supplier Number: CS-P0130010C
  • 10 ml
    Peptide synthesis vessel, solid phase, T-bore PTFE stopcock
    14
    15 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $328.93
    $328.93
    Case of 1
       $328.93
    Supplier Number: CS-P0130010M
  • 025 ml
    Peptide synthesis vessel, solid phase, T-bore PTFE stopcock
    25
    20 mm
    Coarse
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $364.93
    $364.93
    Case of 1
       $364.93
    Supplier Number: CS-P0130025C
  • 25 ml
    Peptide synthesis vessel, solid phase, T-bore PTFE stopcock
    25
    20 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $364.93
    $364.93
    Case of 1
       $364.93
    Supplier Number: CS-P0130025M
  • 050 ml
    Peptide synthesis vessel, solid phase, T-bore PTFE stopcock
    25
    25 mm
    Coarse
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $395.78
    $395.78
    Case of 1
       $395.78
    Supplier Number: CS-P0130050C
  • 50 ml
    Peptide synthesis vessel, solid phase, T-bore PTFE stopcock
    25
    25 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $395.78
    $395.78
    Case of 1
       $395.78
    Supplier Number: CS-P0130050M
  • 100 ml
    Peptide synthesis vessel, solid phase, T-bore PTFE stopcock
    25
    40 mm
    Coarse
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $430.21
    $430.21
    Case of 1
       $430.21
    Supplier Number: CS-P0130100C
  • 100 ml
    Peptide synthesis vessel, solid phase, T-bore PTFE stopcock
    25
    40 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $430.21
    $430.21
    Case of 1
       $430.21
    Supplier Number: CS-P0130100M
  • 250 ml
    Peptide synthesis vessel, solid phase, T-bore PTFE stopcock
    32
    50 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $547.40
    $547.40
    Case of 1
       $547.40
    Supplier Number: CS-P0130250M