Peptide Synthesis Vessels, Solid Phase, Chemscience
Supplier: CHEM SCIENCE INC CA
Jump to:
À propos de cet article
With a fritted disc of coarse or medium porosity to support resin.
- Top of the vessel has GL thread and PTFE lined screw cap
PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
9 Options Available Below
Product Details & Documents
With a fritted disc of coarse or medium porosity to support resin.
- Top of the vessel has GL thread and PTFE lined screw cap
PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Product Family Options
Informations sur le produit
- CapacitéDésignationGL Thread SizeFritted Disc SizePorositéAvailabilityPrice
- 10 mlPeptide synthesis vessel, solid phase1415 mmCoarseLog in pour la disponibilitéCase of 1$258.24Supplier Number: CS-P0120010C
- 10 mlPeptide synthesis vessel, solid phase1415 mmMediumLog in pour la disponibilitéCase of 1$258.24Supplier Number: CS-P0120010M
- 25 mlPeptide synthesis vessel, solid phase2520 mmCoarseLog in pour la disponibilitéCase of 1$293.56Supplier Number: CS-P0120025C
- 25 mlPeptide synthesis vessel, solid phase2520 mmMediumLog in pour la disponibilitéCase of 1$293.56Supplier Number: CS-P0120025M
- 50 mlPeptide synthesis vessel, solid phase2525 mmCoarseLog in pour la disponibilitéCase of 1$327.62Supplier Number: CS-P0120050C
- 50 mlPeptide synthesis vessel, solid phase2525 mmMediumLog in pour la disponibilitéCase of 1$327.62Supplier Number: CS-P0120050M
- 100 mlPeptide synthesis vessel, solid phase2540 mmCoarseLog in pour la disponibilitéCase of 1$424.97Supplier Number: CS-P0120100C
- 100 mlPeptide synthesis vessel, solid phase2540 mmMediumLog in pour la disponibilitéCase of 1$424.97Supplier Number: CS-P0120100M
- 250 mlPeptide synthesis vessel, solid phase3250 mmMediumLog in pour la disponibilitéCase of 1$465.43Supplier Number: CS-P0120250M