Vacuum Manifolds, Single Body, Chemscience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
These manifolds have four high vacuum solid glass stopcocks of 4 mm bore and inner joints.
- T-bore high vacuum stopcock at the inlet end of the main tube
- Main tube has 22 mm O.D.
All glass stopcocks have constant pressure retention devices.
Frequently Bought Together