KIMBLE® Gas Sampling Tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
About this item
These gas sampling tubes with PTFE valves are designed for the collection and storage of environmental samples.
- High vacuum valves are provided for evacuation and filling
- Open top cap with a PTFE/silicone septum allows analytical samples to be withdrawn
- Both ends are ground to accept ³/₈" ferrules and hand-compressible fittings for a gas-tight seal
- Each size has a marking area for easy identification
Valve plug is KT826601-0004. Replacement septum is KT774161-0013.
: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
3 Options Available Below
Product Details & Documents
These gas sampling tubes with PTFE valves are designed for the collection and storage of environmental samples.
- High vacuum valves are provided for evacuation and filling
- Open top cap with a PTFE/silicone septum allows analytical samples to be withdrawn
- Both ends are ground to accept ³/₈" ferrules and hand-compressible fittings for a gas-tight seal
- Each size has a marking area for easy identification
Valve plug is KT826601-0004. Replacement septum is KT774161-0013.
: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Product Family Options
Product Information
- CapacitéDésignationLongueurAvailabilityPrice
- Supplier Number: 653150-0125
- Supplier Number: 653150-0250
- Supplier Number: 653150-0500