KIMBLE® Separatory Funnels, Squibb Type, DWK Life Sciences
À propos de cet article
KONTES® Squibb separatory funnels with PTFE stopcock and standard taper joint.
- Plain stem, but may be used with PTFE BEVEL-SEAL™ inlet adapter (Note stem diameter for a proper fit when using this adapter)
- With an 1:5 taper PTFE stopcock plug
- Manufactured from 33 expansion, low extractable borosilicate glass
KT636031-1000 is supplied without a stopper. KIMBLE® separatory funnels, squibb type (KT636030 series) is supplied with a glass stopper.
: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
8 Options Available Below
Product Details & Documents
KONTES® Squibb separatory funnels with PTFE stopcock and standard taper joint.
- Plain stem, but may be used with PTFE BEVEL-SEAL™ inlet adapter (Note stem diameter for a proper fit when using this adapter)
- With an 1:5 taper PTFE stopcock plug
- Manufactured from 33 expansion, low extractable borosilicate glass
KT636031-1000 is supplied without a stopper. KIMBLE® separatory funnels, squibb type (KT636030 series) is supplied with a glass stopper.
: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Certificates
Recherche par numéro de lotProduct Family Options
Informations sur le produit
- DésignationCapacitéHauteurAvailabilityPrice
- Supplier Number: 636030-0060
- Supplier Number: 636030-0125
- Supplier Number: 636030-0250
- Supplier Number: 636030-0500
- Supplier Number: 636030-1000
- Supplier Number: 636031-1000
- Supplier Number: 636030-2000
- Seporatory funnel4000 ml542 mmLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,064.70$1,064.70Prix promotionnel pour les nouveaux clientsEach$1,064.70Supplier Number: 636030-4000