KIMBLE® Reaction Flasks, Cylindrical, Jacketed, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Jacketed cylindrical reaction flasks with a flat ground flange and a ³/₁₆" wall construction.
- Manufactured from 33 expansion, low extractable borosilicate glass
This 4" cylindrical reaction flasks accept PTFE blade KT789030-0022 and clamp 10135-804.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together