KIMBLE® Distilling Flasks, Jacketed, DWK Life Sciences
500 ml, Distillation flask, 24/40 center; 24/40 side
Catalog # KT609500-0524
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 609500-0524
KIMBLE® Distilling Flasks, Jacketed, DWK Life Sciences
Catalog # KT609500-0524
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
Jacketed three-neck round-bottom reaction flask with standard taper outer joints, all vertical.
- Heavy wall
- Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Spécifications
- Capacité:500 ml
- Désignation:Distillation flask
- ST Joint Size:24/40 center; 24/40 side