Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
BBL™ Salt Broth, Modified, BD

BBL™ Salt Broth, Modified, BD

Supplier:   BD
BBL™ Salt Broth, Modified, BD
Supplier:  BD
Certificates

1 Options Available Below

Product Details & Documents

BBL™ Salt Broth (Modified) is a selective medium for the differentiation of Enterococcus species from the Streptococcus bovis group of streptococci.

  • Growth (turbidity) within 72 hours indicates that the strain is salt tolerant
  • Sometimes changes the indicator color from purple to yellow within 72 hours
  • Broth media containing 6.5% NaCl are used to differentiate enterococci by determining the salt tolerance

Digests of animal tissue provide nitrogen and carbon compounds essential for bacterial growth. Dextrose is an energy source.

Bromcresol purple is an indicator of acid production during bacterial metabolism. Sodium chloride serves as a selective and differential agent.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CA-90004-756
  • CA-90004-582
  • Storage conditions:
  • 2...25 °C
  • 2...25 °C

Specifications

Product Family Options

Product Information

  • Désignation
    Taille
    Tube Size
    Availability
    Price
  • BBL™ salt broth, modified
    100×2 ml
    Size K
    Log in for availability
    Log in for availability
    Pack of 100 (2ml)
    Pack of 100 (2ml) - $750.00
    $750.00
    Pack of 100 (2ml)
       $750.00
    Minimum order quantity is 1

    Supplier Number: 297189
    Specifications:
    More
    Specifications
    Cat. No.
    CA-90004-756
    Storage conditions
    2...25 °C
  • BBL™ salt broth, modified
    100×10 ml
    Size D
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    CA-90004-582
    Storage conditions
    2...25 °C