KIMBLE® Distilling Flask, Pear Shaped with Side Tubulation, DWK Life Sciences
Catalog # KT294500-0010
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:10 ml
- Description:Distillation flask
- Environmentally Preferable:
- Cat. no.:KT294500-0010
- ST Joint Size:14/20
- Supplier No.:294500-0010
Specifications
About this item
Pear-shaped flask with a standard taper outer joint and 7 mm ID side tabulation.
- Heavy wall
- Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.