VWR® 4-Leg Lab Benches
Fournisseur: VWR International
Certificats
À propos de cet article
The classically designed VWR® four-leg lab bench is the ideal basic bench, the simplest and the most cost-effective option in range of workbenches.
- Steel table frame, epoxy powder coated light neutral
- Supports up to 2000 lbs.
- Easily add a variety of accessories to create a complete work space
- Height adjustable with an allen key between 27.56 to 40.55"
- Phenolic and laminate worksurfaces available
Frame color is light neutral. Top features black phenolic or black laminate options. Laminate tops are 1.1875" thick, phenolic tops are 0.75" thick.
Informations sur les accessoires: Additional accessories are available.
Delivery: Assembly is required. Tables are delivered unassembled, unless otherwise ordered.
Frequently Bought Together