Empore™ Solid Phase Extraction Disks, CDS Analytical
À propos de cet article
Empore SPE disks are a cost-effective and trusted solution in large-volume sample processing for more than 30 years.
- Large-volume processing – Ultra-fast flow up to 700 ml/min can be achieved
- Avoid fine particulates – Particulates cause clogging on column frits, which is common with glass-fiber disks; with the Empore disk, sorbent is tightly bound in a polymer matrix eliminating the shedding of fine particulates
- EPA compliant with drinking and wastewater testing methods
- Improved extraction efficiency – High density sorbent bed improves interactions between sorbent particles and analyte
- Excellent reproducibility – Uniform particle bed improves lot-to-lot consistency
Empore SPE disks are EPA compliant, have excellent reproducibility, are optimized for fast flow for large volume samples, and do not contribute to degradation of column life due to the unique nature of the Empore membrane.
15 Options Available Below
Product Details & Documents
Empore SPE disks are a cost-effective and trusted solution in large-volume sample processing for more than 30 years.
- Large-volume processing – Ultra-fast flow up to 700 ml/min can be achieved
- Avoid fine particulates – Particulates cause clogging on column frits, which is common with glass-fiber disks; with the Empore disk, sorbent is tightly bound in a polymer matrix eliminating the shedding of fine particulates
- EPA compliant with drinking and wastewater testing methods
- Improved extraction efficiency – High density sorbent bed improves interactions between sorbent particles and analyte
- Excellent reproducibility – Uniform particle bed improves lot-to-lot consistency
Empore SPE disks are EPA compliant, have excellent reproducibility, are optimized for fast flow for large volume samples, and do not contribute to degradation of column life due to the unique nature of the Empore membrane.
Documents
Certificates
Recherche par numéro de lotProduct Family Options
Informations sur le produit
- DésignationØÉpaisseurTaille de poreAvailabilityPrice
- Empore™ SPE disk, oil and grease47 mm50 µmPack of 20$210.03Case of 60$630.09Supplier Number: 98-0405-0062-5
- Empore™ SPE disk, C847 mm0.5 mm12 µmPack of 20$406.39Case of 60$1,219.17Supplier Number: 98-0604-0214-0
- Empore™ SPE disk, C1847 mm0.5 mm12 µmPack of 20$348.63Case of 60$1,045.89Supplier Number: 98-0604-0217-3
- Empore™ SPE disk, SDB-XC47 mm0.5 mm12 µmPack of 20$406.94Case of 60$1,220.82Supplier Number: 98-0604-0223-1
- Empore™ SPE disk, SDB-RPS47 mm0.5 mm12 µmPack of 20$621.43Case of 60$1,864.29Supplier Number: 98-0604-0226-4
- Empore™ SPE disk, strong anion exchange47 mm0.5 mm12 µmPack of 20$448.23Case of 60$1,344.69Supplier Number: 98-0604-0229-8
- Empore™ SPE disk, strong cation exchange47 mm0.5 mm12 µmPack of 20$564.94Case of 60$1,694.82Supplier Number: 98-0604-0232-2
- Empore™ SPE disk, carbon47 mm0.5 mm12 µmPack of 20$620.79Case of 60$1,862.37Supplier Number: 98-0604-0235-5
- Empore™ SPE disk, chelating47 mm0.5 mm12 µmDiscontinued-Supplier Number: -
- Empore™ SPE disk, SDB-XD47 mmPack of 20$630.91Case of 60$1,892.73Supplier Number: 98-0604-0240-5
- Empore™ SPE disk, oil and grease90 mm50 µmPack of 10$206.06Case of 30$618.18Supplier Number: 98-0405-0063-3
- Empore™ SPE disk, C890 mm0.5 mm12 µmPack of 10$737.18Case of 30$2,211.54Supplier Number: 98-0604-0215-7
- Empore™ SPE disk, C1890 mm0.5 mm12 µmPack of 10$706.52Case of 30$2,119.56Supplier Number: 98-0604-0218-1
- Empore™ SPE disk, SDB-XC90 mm0.5 mm12 µmPack of 10$1,001.97Case of 30$3,005.91Supplier Number: 98-0604-0224-9
- Empore™ SPE disk, SDB-RPS90 mm0.5 mm12 µmDiscontinued-Supplier Number: -
- Empore™ SPE disk, strong anion exchange90 mm0.5 mm12 µmPack of 10$1,071.68Case of 30$3,215.04Supplier Number: 98-0604-0230-6
- Empore™ SPE disk, carbon90 mm0.5 mm12 µmPack of 10$1,160.60Case of 30$3,481.80Supplier Number: 98-0604-0236-3