Empore™ Solid Phase Extraction Cartridges, CDS Analytical
Empore™ Solid Phase Extraction Cartridges, CDS Analytical
Supplier:   CDS ANALYTICAL
Empore™ Solid Phase Extraction Cartridges, CDS Analytical
Supplier:  CDS ANALYTICAL
Certificates
Empore™ Solid Phase Extraction Cartridges, CDS Analytical
Empore™ Solid Phase Extraction Cartridges, CDS Analytical
4 Options Available Below
Product Details & Documents
Improve quality of data and save time and money with Empore SPE cartridges.
  • Eliminate evaporation steps with the lowest sample and elution volumes
  • Avoid clogging of frits, commonly caused by loose-packed SPE cartridges, to reduce LC-MS downtime
  • Improved extraction efficiency – High density sorbent bed improves interactions between sorbent particles and analyte
  • High reproducibility – Uniform particle bed improves lot-to-lot consistency
  • Universal compatibility – Cartridge design features a standard luer tip fitting compatible with standard cartridge manifolds
The unique matrix of the Empore membrane allows for optimized elution volume, which improves sensitivity, reproducibility, provides cleaner chromatograms, and eliminates evaporation time to improve throughput. Additionally, the Empore membrane is resistant to clogging and will not contribute to degradation of instrument column lifetime.
Product Family Options
Product Information
  • Désignation
    Granulométrie
    Ø ext.
    Capacité
    Bed Weight
    Availability
    Price
  • Empore™ SPE disk cartridge, C8-HD, 0.5 mm thick
    12 µm
    4 mm
    1 ml
    4 mg (loose-packed sorbent equivalent 50 mg)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $995.72
    $995.72
    Log in for availability
    Case of 300
    Case of 300 - $2,987.16
    $2,987.16
    Pack of 100
       $995.72
    Case of 300
       $2,987.16
    Supplier Number: 98-0604-0188-6
  • Empore™ SPE disk cartridge, C8-SD, 0.75 mm thick
    50 µm
    4 mm
    1 ml
    5.5 mg (loose-packed sorbent equivalent 50 mg)
    Discontinued
    Supplier Number: -
  • Empore™ SPE disk cartridge, C18-SD, 0.75 mm thick
    50 µm
    4 mm
    1 ml
    5.5 mg (loose-packed sorbent equivalent 50 mg)
    Discontinued
    Supplier Number: -
  • Empore™ SPE disk cartridge, C18-SD, 0.75 mm thick
    50 µm
    7 mm
    3 ml
    17 mg (loose-packed sorbent equivalent 100 mg)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $708.30
    $708.30
    Log in for availability
    Case of 150
    Case of 150 - $2,124.90
    $2,124.90
    Pack of 50
       $708.30
    Case of 150
       $2,124.90
    Supplier Number: 98-0604-0198-5
  • Empore™ SPE disk cartridge, C18-SD, 0.75 mm thick
    50 µm
    10 mm
    6 ml
    34 mg (loose-packed sorbent equivalent 200 mg)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $662.89
    $662.89
    Log in for availability
    Case of 90
    Case of 90 - $1,988.67
    $1,988.67
    Pack of 30
       $662.89
    Case of 90
       $1,988.67
    Supplier Number: 98-0604-0199-3
  • Empore™ SPE disk cartridge, SDB-XC, 0.5 mm thick
    12 µm
    10 mm
    6 ml
    15 mg (loose-packed sorbent equivalent 200 mg)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $335.46
    $335.46
    Log in for availability
    Case of 90
    Case of 90 - $1,006.38
    $1,006.38
    Pack of 30
       $335.46
    Case of 90
       $1,006.38
    Supplier Number: 98-0604-0203-3