Empore™ Solid Phase Extraction RAD Disks, CDS Analytical | Avantor
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
Supplier:   CDS ANALYTICAL
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
Supplier:  CDS ANALYTICAL
Certificates
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
3 Options Available Below
Product Details & Documents
Empore RAD disks provide an efficient alternative to conventional radiochemical sample preparation methods that use wet chemistry or packed columns.
  • EPA compliant with radiochemical testing methods
  • Improved safety by reducing usage of concentrated acids needed for precipitation methods
  • Easier sample handling and waste management by avoiding excessively large sample volumes needed for other radiochemical testing methods
  • Large-volume processing – Ultra-fast flow up to 700 mL/min can be achieved
  • Improved extraction efficiency – High density sorbent bed improves interactions between sorbent particles and analyte
  • Excellent reproducibility – Uniform particle bed improves lot-to-lot consistency
Empore RAD SPE disks trap radiochemical species by molecular recognition technology. Concentration, separation, and sample counting preparation are all easily combined into a single step.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Épaisseur
    Availability
    Price
  • Empore™ RAD disk, Strontium (Sr-90)
    47 mm
    0.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $4,680.85
    $4,680.85
    Log in pour la disponibilité
    Case of 60
    Case of 60 - $14,042.55
    $14,042.55
    Pack of 20
       $4,680.85
    Case of 60
       $14,042.55
    Supplier Number: 98-0405-0064-1
  • Empore™ RAD Disk, Radium (Ra-226, Ra-228)
    47 mm
    0.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $4,991.85
    $4,991.85
    Log in pour la disponibilité
    Case of 60
    Case of 60 - $14,975.55
    $14,975.55
    Pack of 20
       $4,991.85
    Case of 60
       $14,975.55
    Supplier Number: 98-0405-0065-8
  • Empore™ RAD Disk, Technetium (Tc-99)
    47 mm
    0.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $999.59
    $999.59
    Log in pour la disponibilité
    Case of 60
    Case of 60 - $2,998.77
    $2,998.77
    Pack of 20
       $999.59
    Case of 60
       $2,998.77
    Supplier Number: 98-0405-0066-6