Empore™ 96-Well Solid Phase Extraction Plates, CDS Analytical
Supplier:   CDS ANALYTICAL
Empore™ 96-Well Solid Phase Extraction Plates, CDS Analytical
Supplier:  CDS ANALYTICAL
Certificates
Empore™ 96-Well Solid Phase Extraction Plates, CDS Analytical
Empore™ 96-Well Solid Phase Extraction Plates, CDS Analytical
5 Options Available Below
Product Details & Documents
Empore SPE 96-well plates improve data and throughput at a low cost.
  • Optimized for smaller sample volumes, which in turn, reduces elution volumes.
  • Eliminate evaporation steps to streamline methods, improve throughput, and reduce labor.
  • Universal compatibility – Standard polypropylene 96-well plate design for automated applications
  • Avoid clogging of frits by fine particulates, which improves LC-MS uptime and column life
  • Excellent reproducibility – Uniform particle bed improves lot-to-lot consistency
The unique matrix of the Empore membrane allows for optimized elution volume, which improves sensitivity, reproducibility, and provides cleaner chromatograms. Additionally, using less solvent for elution shortens or eliminates evaporation time which improves throughput.
Diameter: 5.5 mm
Product Family Options
Informations sur le produit
  • Désignation
    Volume
    Availability
    Price
  • Standard Empore™ 96-well filter plate
    1.2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $133.79
    $133.79
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,605.48
    $1,605.48
    Each
       $133.79
    Case of 12
       $1,605.48
    Supplier Number: 6065
  • Standard Empore™ 96-well plate, C8-SD, 12 µm, 0.75 mm thick
    1.2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $516.18
    $516.18
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $6,194.16
    $6,194.16
    Each
       $516.18
    Case of 12
       $6,194.16
    Supplier Number: 70-2007-3981-4
  • Standard Empore™ 96-well plate, C18-SD, 12 µm, 0.75 mm thick
    1.2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $516.18
    $516.18
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $6,194.16
    $6,194.16
    Each
       $516.18
    Case of 12
       $6,194.16
    Supplier Number: 70-2007-3982-2
  • Deep Empore™ 96-well filter plate
    2.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $138.11
    $138.11
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,657.32
    $1,657.32
    Each
       $138.11
    Case of 12
       $1,657.32
    Supplier Number: 70-2007-3988-9
  • Standard Empore™ 96-well plate, Mixed Phase Cation (MPC), 32 µm, 0.75 mm thick
    1.2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $516.18
    $516.18
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $6,194.16
    $6,194.16
    Each
       $516.18
    Case of 12
       $6,194.16
    Supplier Number: 70-2007-3983-0