Compartment Tables
Compartment Tables
Compartment Tables
These book compartment tables are built with the same quality techniques as the plain apron table. These tables have either two or four book compartments (depending on size) installed in the apron.
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Finished natural with a chemical resistant UV finish
  • Minimal assembly required; legs need to be attached
  • Solid oak construction
Book compartments measureW×D×H: 14×13⁵/₈×3⁷/₈"
Features 2¼" solid square legs, with ⁵/₁₆" hanger bolts installed, supplied nuts and washers and ¾" phenolic table surface.
Aprons are ¹³/₁₆×4½" with heavy-duty 13 gauge steel corner braces.
 : SEFA compliant and MAS Certified Green.
 : On tables 72"W or over, leg braces and stretchers is recommended to be ordered separately.
  • Catalog No:
  • 470117-854
  • 470117-856
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • MAS Certified Green
  • MAS Certified Green
Product Family Options
  • Désignation
    Capacité
    Worksurface Color/Finish
    Worksurface Material
    Worksurface Thickness
    Base/Leg Material
    Hauteur
    W×D
  • Book compartment table
    500 lbs.
    Black
    Phenolic resin
    2¼"
    Oak
    30"
    60×24"
    Each
    Each - $1,862.65
    $1,862.65
    Each
       $1,862.65
     : C7604K30N
      Safer Human and Environmental Health
    Specifications
    Cat. No.
    470117-854
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS Certified Green
      Safer Human and Environmental Health
  • Book compartment table
    500 lbs.
    Black
    Phenolic resin
    2¼"
    Oak
    36"
    60×24"
    Each
    Each - $2,004.85
    $2,004.85
    Each
       $2,004.85
     : C7604K36N
      Safer Human and Environmental Health
    Specifications
    Cat. No.
    470117-856
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS Certified Green
      Safer Human and Environmental Health