KONTES® Quartz Beaker, Kimble®
Catalog # KT319011-0050
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:50 ml
- Description:Quartz Beaker Only
- Cat. no.:KT319011-0050
- Supplier No.:319011-0050
Specifications
About this item
This quartz beaker/crucible is designed for muffle furnace ashing procedures.
- Useful where chemical purity is a factor in subsequent analysis
- Quartz will also satisfy high temperature requirements to 1050°C
- Very useful for determining trace metals in environmental samples
The cover has to be ordered separately.