VWR® Solid Oak Two Student Tables with Cabinet | Avantor
VWR® Solid Oak Two Student Tables with Cabinet
   Options durables disponibles
Supplier:   VWR International
VWR® Solid Oak Two Student Tables with Cabinet
Supplier:  VWR International
   Options durables disponibles
Certificates
VWR® Solid Oak Two Student Tables with Cabinet
3 Options Available Below
Product Details & Documents
Two student cabinet table constructed of solid oak, oak and hardwood veneers.
  • Cabinet base unit with full door, has an adjustable shelf with a 40 lbs. capacity per sq. foot
  • 1¼" black ChemGuard surface
  • Finishing the desk is rubber base molding, leg boots and a durable chemical resistant UV finish
  • This cabinet table allows workspace and storage for two users
This table is available with plain or book compartment aprons and comes in both single (two-student) and double faced (four-student) models. The center cabinet is available with two locking drawers or as a plain base cabinet with adjustable shelf. These tables are designed for use at sitting height. They are finished with a chemical resistant, earth-friendly UV finish.
Certifications: It is SEFA compliant and MAS certified green.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89374-860
  • 89374-876
  • 89374-868
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • MAS Certified Green
  • MAS Certified Green
  • MAS Certified Green
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    No. of Users
    Worksurface Material
    Hauteur
    W×D
    Availability
    Price
  • Table with cabinet
    2
    ChemGuard
    30"
    66×24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,562.59
    $5,562.59
    Each
       $5,562.59
    La quantité minimale de commande est de 1
    Spécifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    89374-860
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS Certified Green
      Safer Human and Environmental Health
  • Table with cabinet
    2
    Epoxy resin
    30"
    66×24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,295.64
    $7,295.64
    Each
       $7,295.64
    La quantité minimale de commande est de 1
    Spécifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    89374-876
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS Certified Green
      Safer Human and Environmental Health
  • Table with cabinet
    2
    Phenolic resin
    30"
    66×24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,611.91
    $6,611.91
    Each
       $6,611.91
    La quantité minimale de commande est de 1
    Spécifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    89374-868
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS Certified Green
      Safer Human and Environmental Health