VWR® Four Student Tables with Cabinet Made from Solid Oak
Four student cabinet table constructed of solid oak, oak and hardwood veneers.
- 1¼" ChemGuard table surface
- Four student workstation features two locking center cabinets with four locking drawers
- Includes four book compartments in the aprons
- The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
This table features a center cabinet with lock, and rubber base molding. This table is available with plain or book compartment aprons and comes in both single (two-student) and double faced (four-student) models. These tables are designed for use at sitting height.
: It is SEFA compliant and MAS certified green.
- Catalog No:
- 89374-898
- 89374-906
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
Frequently Bought Together