Medi-Waste Biohazardous Bags, Medegen Medical Products
Medi-waste bags from offer an etra-strength solution for heavy-duty waste containment, transport, and removal.
Made from heavy-duty LLDPE, ranging from 1 ply up to 3 ply formats.
: Dual tested bags meet ASTM Dart Impact and Elmedorf tear testing. DOT CFR 49 173-197 compliant.
: Bags are supplied with a drawstring and clip.
Frequently Bought Together