Chemotherapy Waste Bags, Medegen Medical Products
Chemotherapy waste collection bags are ideal for transport of anti-neoplastic medications and chemotherapy waste.
- Heavy-duty construction for puncture and tear resistance
- Dual tested for use in all 50 states
- Made in the USA
Bags feature reinforced seals for leak resistance and meet all labeling requirements for CYTA collection. All bags are color-coded (yellow film) and feature bold labeling with International Biohazard Symbol for easy identification.
Frequently Bought Together