Anti-Static Kick Bucket Liners, Medegen Medical Products
Anti-static kick bucket liners comply with operating room requirements.
- Bags with size of 12×24×8" fit round kick buckets
- Bags with size of 18×16×8" fit oval kick buckets
- Red film bags feature a biohazard print
- Blue film bags feature a soiled linen print
Liners contain a carbon additive to help prevent the build-up of electrostatic charges.
: Meets NFPA Standard 56A.
Frequently Bought Together