ICERoam35 Mobile Dry Ice Reusable Transport Container
Catalog # 76333-166
Supplier: ROEMER INDUSTRIES, INC.
Specifications
- Capacity:35 L (refrigerated payload)
20.41 kg (45 lbs., Dry Ice, Net) - Description:Mobile Dry Ice Reusable Transport Container
- Cat. no.:76333-166
- Exterior Dimensions:28×25.6×15"
- Weight:34 kg (75 lbs.) (total, with dry ice)
Specifications
About this item
Withstand over 200 trips by air, land and sea with military-grade aluminum.
- Over 3" of injected polyurethane insulation
- 35 liters of storage space
- Water tight, TIG welded aluminum payload compartment tub
With dry ice in container, temperature will hold for three to five days.
Refrigerated Payload Dimension: (L × W × H) 19×17×7"