SuperBlock™ T20 (PBS) Blocking Buffer, Thermo Scientific | Avantor
SuperBlock™ T20 (PBS) Blocking Buffer, Thermo Scientific
PBS with Tween® 20, 1 L, ELISA, Western Blot, Dot Blot, Immunohistochemistry, Hybridization
Catalog # CAPI37516
Supplier:   Thermo Scientific
Fournisseur #:  37516
SuperBlock™ T20 (PBS) Blocking Buffer, Thermo Scientific
Catalog # CAPI37516
Supplier:  Thermo Scientific
CAS Number:  
SuperBlock™ T20 (PBS) Blocking Buffer, Thermo Scientific
SuperBlock™ T20 (PBS) Blocking Buffer, Thermo Scientific
PBS with Tween® 20, 1 L, ELISA, Western Blot, Dot Blot, Immunohistochemistry, Hybridization
CAS Number:  
Price
$661.92
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
SuperBlock™ Blocking Buffer is optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.
  • Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
  • Stability—store buffer at 4 °C for one year; store blocked plates dry for up to 12 months
Blocking micro-plates, membranes or tissues with a SuperBlock Buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.
Spécifications
Spécifications
  • Désignation:
    PBS with Tween® 20
  • Taille:
    1 L
  • Utilisation:
    ELISA, Western Blot, Dot Blot, Immunohistochemistry, Hybridization