ContactUsLinkComponent
Pressure Transducer for Krosflo Pressure Monitor, Spectrum® Laboratories
Pressure Transducer for Krosflo Pressure Monitor, Spectrum® Laboratories
  97004-062
Small Business Enterprise
undefined
Pressure Transducer for Krosflo Pressure Monitor, Spectrum® Laboratories
  97004-062
 :  ACPM-499-03N
Small Business Enterprise
  • Description:
    Disposable Pressure Transducer
  • Cat. no.:
    97004-062
  • Supplier No.:
    ACPM-499-03N
Polycarbonate and polysulfone single-use pressure transducers are designed to accurately measure feed, permeate, and retentate pressure during Tangential Flow Filtration process.