Chromatography Columns, With Top Outer Joint, ChemScience
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
These short columns with lower 24/40 joint and hose connection to apply vacuum.
Column length is measured from bottom of top joint to fritted disc.
Frequently Bought Together