Chromatography Columns, With Reservoir, Spherical Joint, Fritted Disc, ChemScience
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
These columns with reservoir have a Teflon® stopcock and a 35/20 spherical joint.
Frequently Bought Together