Chromatography Columns, Spherical Joint, ChemScience
These columns have a Teflon® stopcock and a spherical joint.
The spherical joint used with pinch clamp makes quick and tight connection under pressure for flash chromatography.
Frequently Bought Together