Schlenk Tubes, Modified, ChemScience
Supplier: CHEM SCIENCE INC CA
Jump to:
À propos de cet article
Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.
38 Options Available Below
Product Details & Documents
Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.
Product Family Options
Informations sur le produit
- CapacitéDésignationST Joint SizeStopcock TypeAvailabilityPrice
- Supplier Number: CS-F0901410
- Supplier Number: CS-F0901425
- Supplier Number: CS-F0901450
- Supplier Number: CS-F0901950
- Supplier Number: CS-F0902450
- Supplier Number: CS-F0901100
- Supplier Number: CS-F0909100
- Supplier Number: CS-F0904100
- Supplier Number: CS-F0901200
- Supplier Number: CS-F0909200
- Supplier Number: CS-F0904200
- Supplier Number: CS-F0904500
- Supplier Number: CS-F0904000
- Supplier Number: CS-F0901410G
- Supplier Number: CS-F0901425G
- Supplier Number: CS-F0901450G
- Supplier Number: CS-F0901950G
- Supplier Number: CS-F0902450G
- Supplier Number: CS-F0901100G
- Supplier Number: CS-F0909100G
- Supplier Number: CS-F0904100G
- Supplier Number: CS-F0901200G
- Supplier Number: CS-F0909200G
- Supplier Number: CS-F0904200G
- Supplier Number: CS-F0904500G
- Supplier Number: CS-F0904000G
- Supplier Number: CS-F0901425H
- Supplier Number: CS-F0901450H
- Supplier Number: CS-F0901950H
- Supplier Number: CS-F0902450H
- Supplier Number: CS-F0901100H
- Supplier Number: CS-F0909100H
- Supplier Number: CS-F0904100H
- Supplier Number: CS-F0901200H
- Supplier Number: CS-F0909200H
- Supplier Number: CS-F0904200H
- Supplier Number: CS-F0904500H
- Supplier Number: CS-F0904000H