Schlenk Tubes, with High Vacuum Valve, ChemScience
These valves have a PTFE wiper to protect Viton® O-ring on shaft.
: Supplied with high vacuum valve with PTFE O-ring at tip.
Frequently Bought Together
These valves have a PTFE wiper to protect Viton® O-ring on shaft.