Full Size Security Units, Eagle MHC | Avantor
Full Size Security Units, Eagle MHC
Supplier:   Eagle Group
Full Size Security Units, Eagle MHC
Supplier:  Eagle Group
Certificates
Full Size Security Units, Eagle MHC
Full Size Security Units, Eagle MHC
9 Options Available Below
Product Details & Documents
Security units are ideal for safely storing and transporting costly materials and items subject to pilferage.
  • Heavy-gauge open-wire construction keeps contents visible for quick inventory checks
  • Variety of sizes are available in stationary models or with casters for easy mobility
  • All units are plated with a durable easy-to-clean finish
  • Quick-action locking feature with hasps for padlock
  • All units feature full-access double door swinging out at approximately 270°
Mobile security units feature 5" (127 mm) diameter swivel stem casters with poly tread, two with brake. Casters on stainless steel units are stainless steel. Casters on chrome and EAGLEgard® units are nickel-plated.
Packaging: Shipped knocked down for easy assembly and reduced transportation costs.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Largeur
    Hauteur
    Weight
    Availability
    Price
  • Chrome Stationary, Full-Size Security Unit
    63¼"
    33¼"
    67"
    93.9 kg (207 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,911.44
    $6,911.44
    Each
       $6,911.44
    Supplier Number: SC3060
  • Stainless Steel Stationary, Full-Size Security Unit
    51¼"
    27¼"
    67"
    77.6 kg (171 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13,800.42
    $13,800.42
    Each
       $13,800.42
    Supplier Number: SC2448S
  • Chrome Stationary, Full-Size Security Unit
    51¼"
    27¼
    67"
    77.6 kg (171 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,096.97
    $5,096.97
    Each
       $5,096.97
    Supplier Number: SC2448
  • Chrome Stationary, Full-Size Security Unit
    39¼"
    17¼"
    67"
    55.8 kg (123 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,837.52
    $3,837.52
    Each
       $3,837.52
    Supplier Number: SC1436
  • Chrome Dolly Truck, Full-Size Security Unit
    52⅞"
    29¾"
    69"
    97.1 kg (214 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,849.57
    $6,849.57
    Each
       $6,849.57
    Supplier Number: DTSC2448
  • Chrome Mobile, Full-Size Security Unit. Includes Four Poly Stem Swivel Casters 5×11/4", Two With Brakes
    63¼"
    33¼"
    69"
    100.2 kg (221 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,877.95
    $7,877.95
    Each
       $7,877.95
    Supplier Number: CSC3060
  • Chrome Mobile, Full-Size Security Unit. Includes Four Poly Stem Swivel Casters 5×11/4", Two With Brakes
    63¼"
    27¼"
    69"
    92.5 kg (204 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,249.39
    $6,249.39
    Each
       $6,249.39
    Supplier Number: CSC2460
  • Stainless Steel Mobile, Full-Size Security Unit. Includes Four Poly Stem Swivel Casters 5×11/4", Two With Brakes
    51¼"
    27¼"
    69"
    83.9 kg (185 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13,806.05
    $13,806.05
    Each
       $13,806.05
    Supplier Number: CSC2448S
  • Chrome Mobile, Full-Size Security Unit. Includes Four Poly Stem Swivel Casters 5×11/4", Two With Brakes
    51¼"
    27¼"
    69"
    83.9 kg (185 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,812.71
    $5,812.71
    Each
       $5,812.71
    Supplier Number: CSC2448
Accessories
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-004
    36L×14" W
    3.6 kg (8 lbs.)
    Log in pour la disponibilité
    Each - $226.94
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-006
    48L×14" W
    4.5 kg (10 lbs.)
    Log in pour la disponibilité
    Each - $258.02
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-024
    30L×24" W
    5.0 kg (11 lbs.)
    Log in pour la disponibilité
    Each - $289.32
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-026
    36L×24" W
    5.9 kg (13 lbs.)
    Log in pour la disponibilité
    Each - $297.85
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-030
    48L×24" W
    7.3 kg (16 lbs.)
    Log in pour la disponibilité
    Each - $354.59
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-036
    36L×30" W
    7.7 kg (17 lbs.)
    Log in pour la disponibilité
    Each - $436.80
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-040
    60L×30" W
    11.4 kg (25 lbs.)
    Log in pour la disponibilité
    Each - $553.11
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-038
    48L×30" W
    9.1 kg (20 lbs.)
    Log in pour la disponibilité
    Each - $499.36
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-068
    48L×24" W
    7.3 kg (16 lbs.)
    Log in pour la disponibilité
    Each - $1,694.84
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-070
    60L×24" W
    9.5 kg (21 lbs.)
    Log in pour la disponibilité
    Each - $1,653.61
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Eaglegard® Hybrid Epoxy with Microgard®, Wire Shelf
    Eaglegard® Hybrid Epoxy with Microgard®, Wire Shelf
    58202-082
    48L×24" W
    7.3 kg (16 lbs.)
    Log in pour la disponibilité
    Each - $436.80
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-002
    30L×14" W
    3.2 kg (7 lbs.)
    Log in pour la disponibilité
    Each - $212.80
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-920
    30L×14" W
    3.2 kg (7 lbs.)
    Log in pour la disponibilité
    Each - $670.60
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-924
    60L×14" W
    6.4 kg (14 lbs.)
    Log in pour la disponibilité
    Each - $273.30
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-934
    30L×24" W
    5.0 kg (11 lbs.)
    Log in pour la disponibilité
    Each - $1,022.80
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-936
    36L×30" W
    7.7 kg (17 lbs.)
    Log in pour la disponibilité
    Each - $1,372.17
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-938
    48L×30" W
    9.1 kg (20 lbs.)
    Log in pour la disponibilité
    Each - $1,600.42
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-940
    60L×30" W
    11.4 kg (25 lbs.)
    Log in pour la disponibilité
    Each - $1,775.10
Informations sur le produit
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-032
    60L×24" W
    9.5 kg (21 lbs.)
    Log in pour la disponibilité
    Each - $422.67