Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
Supplier:   CHEM SCIENCE INC CA
Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
Certificates
Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
16 Options Available Below
Product Details & Documents
Flasks feature a pear shape with single neck.
  • Heavy wall design
Product Family Options
Product Information
  • Capacité
    ST Joint Size
    Availability
    Price
  • 5 ml
    14/20
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $94.43
    $94.43
    Case of 1
       $94.43
    Supplier Number: CS-F0321405
  • 5 ml
    19/22
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $96.86
    $96.86
    Case of 1
       $96.86
    Supplier Number: CS-F0329205
  • 10 ml
    14/20
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $94.43
    $94.43
    Case of 1
       $94.43
    Supplier Number: CS-F0321410
  • 10 ml
    19/22
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $96.86
    $96.86
    Case of 1
       $96.86
    Supplier Number: CS-F0329210
  • 15 ml
    14/20
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $94.43
    $94.43
    Case of 1
       $94.43
    Supplier Number: CS-F0321415
  • 25 ml
    14/20
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $47.22
    $47.22
    Case of 1
       $47.22
    Supplier Number: CS-F0321225
  • 25 ml
    19/22
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $48.45
    $48.45
    Case of 1
       $48.45
    Supplier Number: CS-F0329225
  • 50 ml
    14/20
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $47.22
    $47.22
    Case of 1
       $47.22
    Supplier Number: CS-F0321250
  • 50 ml
    19/22
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $48.45
    $48.45
    Case of 1
       $48.45
    Supplier Number: CS-F0329250
  • 50 ml
    24/40
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $65.05
    $65.05
    Case of 1
       $65.05
    Supplier Number: CS-F0322450
  • 100 ml
    14/20
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $50.38
    $50.38
    Case of 1
       $50.38
    Supplier Number: CS-F0321100
  • 100 ml
    19/22
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $52.08
    $52.08
    Case of 1
       $52.08
    Supplier Number: CS-F0329100
  • 100 ml
    24/40
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $68.81
    $68.81
    Case of 1
       $68.81
    Supplier Number: CS-F0324100
  • 200 ml
    14/20
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $81.67
    $81.67
    Case of 1
       $81.67
    Supplier Number: CS-F0321200
  • 200 ml
    19/22
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $86.07
    $86.07
    Case of 1
       $86.07
    Supplier Number: CS-F0329200
  • 200 ml
    24/40
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $91.19
    $91.19
    Case of 1
       $91.19
    Supplier Number: CS-F0324200