Antimony Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml
Frequently Bought Together