Solid Shelving, Eagle MHC | Avantor
Solid Shelving, Eagle MHC
Supplier:   Eagle Group
Solid Shelving, Eagle MHC
Supplier:  Eagle Group
Certificates
Solid Shelving, Eagle MHC
Solid Shelving, Eagle MHC
4 Options Available Below
Product Details & Documents
Solid shelving is hemmed, has a two-fold thickness of heavy gauge steel, and comes standard with a marine edge providing unsurpassed strength and stability.
  • Aluminum corner castings create the locking mechanism to fasten the shelves to the posts
  • Raised marine edge on all four sides retains contents to shelf
  • Makes cleaning easier and helps reduce the spill of shelf contents
  • Leveling feet are provided to help compensate for uneven solid shelves and posts combined to form unit floor surfaces
Numerically-calibrated grooved posts, tapered plastic or aluminum split sleeves and aluminum corner castings combine to make shelf assembly fast and easy. Shelving can be vertically adjusted on 1" (25 mm) increments for the entire post length.
Ordering information: Posts are required and sold separately. Contact your Avantor representative for more information.
Product Family Options
Informations sur le produit
  • Désignation
    Largeur
    Longueur
    Weight
    Availability
    Price
  • Galvanized Solid Shelf
    21"
    36"
    18 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $581.50
    $581.50
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $2,326.00
    $2,326.00
    Each
       $581.50
    Case of 4
       $2,326.00
    Supplier Number: SS2136G
  • Galvanized Solid Shelf
    24"
    36"
    19 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $584.40
    $584.40
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $2,337.60
    $2,337.60
    Each
       $584.40
    Case of 4
       $2,337.60
    Supplier Number: SS2436G
  • Stainless Steel Solid Shelf
    24"
    36"
    19 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,154.51
    $1,154.51
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $4,618.04
    $4,618.04
    Each
       $1,154.51
    Case of 4
       $4,618.04
    Supplier Number: SS2436S
  • Stainless Steel Solid Shelf
    24"
    48"
    24 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,401.27
    $1,401.27
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $5,605.08
    $5,605.08
    Each
       $1,401.27
    Case of 4
       $5,605.08
    Supplier Number: SS2448S