Round-Bottom Flasks with Four Vertical Necks, ChemScience
Round-Bottom Flasks with Four Vertical Necks, ChemScience
Round-Bottom Flasks with Four Vertical Necks, ChemScience
Round-Bottom Flasks with Four Vertical Necks, ChemScience
Round-Bottom Flasks with Four Vertical Necks, ChemScience
With identical vertical side joints.
Product Family Options
  • Capacité
    Désignation
    Center Joint
    Side ST Joint Size
  • 500 ml
    Flask, Round Bottom, 4-Neck Vertical
    14/20
    14/20
    Case of 1
    Case of 1 - $288.17
    $288.17
    Case of 1
       $288.17
     : CS-F0211500
  • 500 ml
    Flask, Round Bottom, 4-Neck Vertical
    19/22
    14/20
    Case of 1
    Case of 1 - $293.70
    $293.70
    Case of 1
       $293.70
     : CS-F0219500
  • 500 ml
    Flask, Round Bottom, 4-Neck Vertical
    24/40
    19/22
    Case of 1
    Case of 1 - $298.96
    $298.96
    Case of 1
       $298.96
     : CS-F0214591
  • 500 ml
    Flask, Round Bottom, 4-Neck Vertical
    24/40
    24/40
    Case of 1
    Case of 1 - $310.53
    $310.53
    Case of 1
       $310.53
     : CS-F0214500
  • 500 ml
    Flask, Round Bottom, 4-Neck Vertical
    29/42
    24/40
    Case of 1
    Case of 1 - $341.37
    $341.37
    Case of 1
       $341.37
     : CS-F0212500
  • 500 ml
    Flask, Round Bottom, 4-Neck Vertical
    29/42
    29/42
    Case of 1
    Case of 1 - $351.15
    $351.15
    Case of 1
       $351.15
     : CS-F0218500
  • 1000 ml
    Flask, Round Bottom, 4-Neck Vertical
    24/40
    19/22
    Case of 1
    Case of 1 - $376.42
    $376.42
    Case of 1
       $376.42
     : CS-F0214091
  • 1000 ml
    Flask, Round Bottom, 4-Neck Vertical
    24/40
    24/40
    Case of 1
    Case of 1 - $387.64
    $387.64
    Case of 1
       $387.64
     : CS-F0214000
  • 1000 ml
    Flask, Round Bottom, 4-Neck Vertical
    29/42
    24/40
    Case of 1
    Case of 1 - $401.03
    $401.03
    Case of 1
       $401.03
     : CS-F021841L
  • 2000 ml
    Flask, Round Bottom, 4-Neck Vertical
    24/40
    24/40
    Case of 1
    Case of 1 - $522.48
    $522.48
    Case of 1
       $522.48
     : CS-F021242L
  • 2000 ml
    Flask, Round Bottom, 4-Neck Vertical
    29/42
    24/40
    Case of 1
    Case of 1 - $525.61
    $525.61
    Case of 1
       $525.61
     : CS-F021842L
  • 2000 ml
    Flask, Round Bottom, 4-Neck Vertical
    34/45
    24/40
    Case of 1
    Case of 1 - $542.20
    $542.20
    Case of 1
       $542.20
     : CS-F021342L