Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Supplier: CHEM SCIENCE INC CA
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Supplier: CHEM SCIENCE INC CA
Certificates
Jump to:
À propos de cet article
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
- An outer diameter between 5 and 7.5 mm
- Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
10 Options Available Below
Product Details & Documents
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
- An outer diameter between 5 and 7.5 mm
- Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
Product Family Options
Informations sur le produit
- CapacitéDésignationJoint SizeAvailabilityPrice
- Supplier Number: CS-F0141425
- Supplier Number: CS-F0141450
- Supplier Number: CS-F0141950
- Supplier Number: CS-F0141100
- Supplier Number: CS-F0149100
- Supplier Number: CS-F0144100
- Supplier Number: CS-F0141250
- Supplier Number: CS-F0149250
- Supplier Number: CS-F0144250
- Supplier Number: CS-F0144500