Spécifications
- Capacity:1200 ml
- Description:Scott® Moisturizing Foam Hand Sanitizer
- Cat. no.:CA76333-316
Spécifications
À propos de cet article
How do employers stop the spread of germs when 72% of employees go to work when they are sick? Start by offering Scott® pro moisturizing foam hand sanitizer in the workplace to help protect other employees and maintain the productivity.
- Antimicrobial foam with 62% ethyl alcohol to help reduce the number of germs on hands
- Effectively kills 99.99% of germs on hands in less than 15 seconds, all without the use of water
- Compatible with automatic skin care dispensers
- No rinse formula contains moisturizers and conditioners to hydrate the skin
- Fresh fragrance
An estimated 98% of workers are affected by minor illnesses each year. Additionally, an estimated 80% of infectious illnesses are spread through hands and surfaces. Help stop the spread of germs and keep employees healthy with Scott® pro moisturizing foam hand sanitizer. It effectively kills 99.99% of germs on hands in less than 15 seconds, all without the use of water.
The clear 62% ethyl alcohol formula dispenses in luxury foam and has a refreshing cucumber fragrance. The proprietary antimicrobial formula contains moisturizers and conditioners to hydrate the skin. In the bulk refill cassette format, this no rinse hand sanitizer is designed to fit in compatible Scott® pro dispensers.